COM448683561 Combi-Blocks, Inc.

Fmoc-2,5-dichloro-l-phenylalanine

  • MDL number MFCD07372021
  • Cas Number : 1260614-80-9
  • Molecular Weight : 456.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8899

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8899
Availability details for
Quantity
  • Availability
  • Add to Cart