COM448683599 Combi-Blocks, Inc.

Guaiacylglycerol-beta-guaiacyl ether

  • MDL number MFCD00060144
  • Cas Number : 7382-59-4
  • Molecular Weight : 320.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8967

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8967
Availability details for
Quantity
  • Availability
  • Add to Cart