COM448683738 Combi-Blocks, Inc.

Isoxazol-5-ylmethyl-methyl-amine

  • MDL number MFCD06738816
  • Cas Number : 401647-20-9
  • Molecular Weight : 112.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9163

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9163
Availability details for
Quantity
  • Availability
  • Add to Cart