COM448683796 Combi-Blocks, Inc.

3-Aminopropane-1-sulfonamide, HCl

  • MDL number MFCD12912951
  • Cas Number : 104458-33-5
  • Molecular Weight : 174.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9293

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9293
Availability details for
Quantity
  • Availability
  • Add to Cart