COM448683879 Combi-Blocks, Inc.

3-Amino-4,N,N-trimethyl-benzenesulfonamide

  • MDL number MFCD00025203
  • Cas Number : 6331-68-6
  • Molecular Weight : 214.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9493

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9493
Availability details for
Quantity
  • Availability
  • Add to Cart