COM448683945 Combi-Blocks, Inc.

2-Chloro-5-(trifluoromethyl)cinnamic acid

  • MDL number MFCD03002852
  • Cas Number : 682805-12-5
  • Molecular Weight : 250.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9633

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9633
Availability details for
Quantity
  • Availability
  • Add to Cart