COM448683960 Combi-Blocks, Inc.

Methyl 2-(4-chloropyrimidin-2-yloxy)benzoate

  • MDL number MFCD12068367
  • Cas Number : 1159822-80-6
  • Molecular Weight : 264.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9662

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9662
Availability details for
Quantity
  • Availability
  • Add to Cart