COM448683966 Combi-Blocks, Inc.

Methyl 2-(5-cyanobenzo[b]thiophen-2-yl)acetate

  • MDL number MFCD09910410
  • Cas Number : 1000340-05-5
  • Molecular Weight : 231.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9677

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9677
Availability details for
Quantity
  • Availability
  • Add to Cart