COM448683972 Combi-Blocks, Inc.

Methyl 2-amino-4-(tetrahydropyran-4-yl)-butyrate hcl

  • MDL number MFCD09800520
  • Cas Number : 1255098-55-5
  • Molecular Weight : 237.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9689

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9689
Availability details for
Quantity
  • Availability
  • Add to Cart