COM448683985 Combi-Blocks, Inc.

Methyl 2-methyl-4-benzyloxy-phenylacetate

  • MDL number MFCD01317810
  • Cas Number : 223406-97-1
  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9717

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9717
Availability details for
Quantity
  • Availability
  • Add to Cart