COM448683987 Combi-Blocks, Inc.

Methyl 2-methylpiperidine-2-carboxylate

  • MDL number MFCD13179033
  • Cas Number : 89115-93-5
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9720

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9720
Availability details for
Quantity
  • Availability
  • Add to Cart