COM448683988 Combi-Blocks, Inc.

Methyl 2-n-boc-2-aminomethyl-3-(4-chloro-phenyl)-propionate

  • MDL number MFCD08061214
  • Cas Number : 886366-43-4
  • Molecular Weight : 327.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9721

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9721
Availability details for
Quantity
  • Availability
  • Add to Cart