COM448683996 Combi-Blocks, Inc.

Methyl 3-(4-cyanophenoxy)benzoate

  • MDL number MFCD11040483
  • Cas Number : 934690-54-7
  • Molecular Weight : 253.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9743

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9743
Availability details for
Quantity
  • Availability
  • Add to Cart