COM448684008 Combi-Blocks, Inc.

Methyl 3-amino-4-cyano-5-methylthiophene-2-carboxylate

  • MDL number MFCD12068370
  • Cas Number : 85092-72-4
  • Molecular Weight : 196.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9771

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9771
Availability details for
Quantity
  • Availability
  • Add to Cart