COM448684027 Combi-Blocks, Inc.

Methyl 4-(2-(benzyloxy)acetamido)-2-hydroxybenzoate

  • MDL number MFCD11501881
  • Cas Number : 1092460-39-3
  • Molecular Weight : 315.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9822

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9822
Availability details for
Quantity
  • Availability
  • Add to Cart