COM448684035 Combi-Blocks, Inc.

Methyl 4-acetamido-3-iodobenzoate

  • MDL number MFCD09998274
  • Cas Number : 190071-23-9
  • Molecular Weight : 319.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9843

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9843
Availability details for
Quantity
  • Availability
  • Add to Cart