COM448684036 Combi-Blocks, Inc.

Methyl 4-acetylamino-2-methylbenzate

  • MDL number MFCD11042857
  • Cas Number : 91133-71-0
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9844

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9844
Availability details for
Quantity
  • Availability
  • Add to Cart