COM448684043 Combi-Blocks, Inc.

2,3,5,6-Tetramethoxypyridine-4-boronic acid pinacol ester

  • MDL number MFCD22887384
  • Cas Number : 2096995-77-4
  • Molecular Weight : 325.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9858

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9858
Availability details for
Quantity
  • Availability
  • Add to Cart