COM448684048 Combi-Blocks, Inc.

3-Aminomethyl-piperidine-1-carboxylic acid benzyl ester-hcl

  • MDL number MFCD11101355
  • Cas Number : 1186663-23-9
  • Molecular Weight : 284.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9880

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9880
Availability details for
Quantity
  • Availability
  • Add to Cart