COM448684059 Combi-Blocks, Inc.

Methyl 4-oxo-1-phenylcyclohexanecarboxylate

  • MDL number MFCD09834757
  • Cas Number : 75945-90-3
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9897

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9897
Availability details for
Quantity
  • Availability
  • Add to Cart