COM448684076 Combi-Blocks, Inc.

Methyl 5-benzoyl-furan-2-carboxylate

  • MDL number MFCD07779120
  • Cas Number : 58972-21-7
  • Molecular Weight : 230.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9931

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9931
Availability details for
Quantity
  • Availability
  • Add to Cart