COM448684078 Combi-Blocks, Inc.

4-(Methylamino)-1-benzylpiperidine DiHCl

  • MDL number MFCD11052326
  • Cas Number : 7006-48-6
  • Molecular Weight : 277.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9934

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9934
Availability details for
Quantity
  • Availability
  • Add to Cart