COM448684114 Combi-Blocks, Inc.

Methyl 6-(1-boc-piperidin-4-yloxy)indole-2-carboxylate

  • MDL number MFCD11053686
  • Cas Number : 287389-12-2
  • Molecular Weight : 374.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9996

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9996
Availability details for
Quantity
  • Availability
  • Add to Cart