COM448684140 Combi-Blocks, Inc.

Potassium {3-[(tert-butoxy)carbonyl]phenyl}trifluoroborate

  • MDL number MFCD27936855
  • Molecular Weight : 284.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?TB-2119

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?TB-2119
Availability details for
Quantity
  • Availability
  • Add to Cart