COM448684149 Combi-Blocks, Inc.

Potassium trifluoro[2-(2-hydroxyethyl)phenyl]boranuide

  • MDL number MFCD11113778
  • Cas Number : 1015082-79-7
  • Molecular Weight : 228.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?TB-2481

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?TB-2481
Availability details for
Quantity
  • Availability
  • Add to Cart