COM448684154 Combi-Blocks, Inc.

Potassium [2-(ethylsulfanyl)phenyl]trifluoroborate

  • MDL number MFCD09993638
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?TB-2635

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?TB-2635
Availability details for
Quantity
  • Availability
  • Add to Cart