COM448684158 Combi-Blocks, Inc.

Potassium trifluoro[3-fluoro-4-(methoxycarbonyl)phenyl]borate

  • MDL number MFCD28122701
  • Molecular Weight : 260.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?TB-2719

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?TB-2719
Availability details for
Quantity
  • Availability
  • Add to Cart