COM448684226 Combi-Blocks, Inc.

Potassium trifluoro[2-fluoro-3-(methylcarbamoyl)phenyl]borate

  • MDL number MFCD28143972
  • Molecular Weight : 259.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?TB-4905

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?TB-4905
Availability details for
Quantity
  • Availability
  • Add to Cart