COM448684229 Combi-Blocks, Inc.

Potassium (4-methoxynaphthalen-1-yl)trifluoroborate

  • MDL number MFCD18605929
  • Molecular Weight : 264.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?TB-5250

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?TB-5250
Availability details for
Quantity
  • Availability
  • Add to Cart