COM448684238 Combi-Blocks, Inc.

Potassium trifluoro(2-hydroxy-3-methylphenyl)boranuide

  • MDL number MFCD09993089
  • Cas Number : 1033783-02-6
  • Molecular Weight : 214.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?TB-8561

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?TB-8561
Availability details for
Quantity
  • Availability
  • Add to Cart