COM448684242 Combi-Blocks, Inc.

Trifluoro({[4-(trifluoromethoxy)phenyl]methyl})boranuide potassium

  • MDL number MFCD18713047
  • Cas Number : 1201331-15-8
  • Molecular Weight : 282.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?TB-8734

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?TB-8734
Availability details for
Quantity
  • Availability
  • Add to Cart