COM448684260 Combi-Blocks, Inc.

Methyl 1-butyl-1,2,3-benzotriazole-5-carboxylate

  • MDL number MFCD22421553
  • Cas Number : 1400644-56-5
  • Molecular Weight : 233.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9020

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9020
Availability details for
Quantity
  • Availability
  • Add to Cart