COM448684264 Combi-Blocks, Inc.

Methyl 4-(cyclopentylamino)-3-nitrobenzoate

  • MDL number MFCD14281680
  • Cas Number : 234750-98-2
  • Molecular Weight : 264.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9024

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9024
Availability details for
Quantity
  • Availability
  • Add to Cart