COM448684276 Combi-Blocks, Inc.

1-Benzyl-1,2,3-benzotriazole-5-carbonitrile

  • MDL number MFCD22549343
  • Cas Number : 1242069-41-5
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9037

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9037
Availability details for
Quantity
  • Availability
  • Add to Cart