COM448684289 Combi-Blocks, Inc.

2-Methyl-1-propyl-1,3-benzodiazole-5-carboxylic acid

  • MDL number MFCD11103016
  • Cas Number : 68740-33-0
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9054

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9054
Availability details for
Quantity
  • Availability
  • Add to Cart