COM448684318 Combi-Blocks, Inc.

Methyl 1-benzyl-2-methyl-1,3-benzodiazole-5-carboxylate

  • MDL number MFCD00544325
  • Cas Number : 299927-11-0
  • Molecular Weight : 280.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9090

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9090
Availability details for
Quantity
  • Availability
  • Add to Cart