COM448684319 Combi-Blocks, Inc.

Methyl 1-benzyl-1,2,3-benzotriazole-5-carboxylate

  • MDL number MFCD15006357
  • Cas Number : 951558-74-0
  • Molecular Weight : 267.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9091

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9091
Availability details for
Quantity
  • Availability
  • Add to Cart