COM448684323 Combi-Blocks, Inc.

Methyl 7-bromo-1H-benzotriazole-5-carboxylate

  • MDL number MFCD20501506
  • Cas Number : 1354758-03-4
  • Molecular Weight : 256.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9096

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9096
Availability details for
Quantity
  • Availability
  • Add to Cart