COM448684332 Combi-Blocks, Inc.

7-Bromo-1H-1,2,3-benzotriazole-5-carbonitrile

  • MDL number MFCD23135963
  • Cas Number : 1423037-37-9
  • Molecular Weight : 223.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9107

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9107
Availability details for
Quantity
  • Availability
  • Add to Cart