COM448684347 Combi-Blocks, Inc.

3-(2-Fluorophenyl)-5-methyl-1,2,4-oxadiazole

  • MDL number MFCD23699515
  • Cas Number : 1426958-50-0
  • Molecular Weight : 178.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9127

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9127
Availability details for
Quantity
  • Availability
  • Add to Cart