COM448684349 Combi-Blocks, Inc.

2,3-Difluoro-6-nitro-N-phenylaniline

  • MDL number MFCD24448833
  • Cas Number : 1393178-31-8
  • Molecular Weight : 250.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9130

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9130
Availability details for
Quantity
  • Availability
  • Add to Cart