COM448684370 Combi-Blocks, Inc.

Methyl 7-bromo-1-ethyl-2-methyl-1,3-benzodiazole-5-carboxylate

  • MDL number MFCD23135965
  • Cas Number : 1423037-29-9
  • Molecular Weight : 297.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9165

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9165
Availability details for
Quantity
  • Availability
  • Add to Cart