COM448684373 Combi-Blocks, Inc.

Ethyl 3,4-diamino-5-bromobenzoate

  • MDL number MFCD23135966
  • Cas Number : 1423037-41-5
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9168

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9168
Availability details for
Quantity
  • Availability
  • Add to Cart