COM448684387 Combi-Blocks, Inc.

7-Bromo-1-isopropylbenzotriazole-5-carbonitrile

  • MDL number MFCD24448836
  • Cas Number : 1437794-89-2
  • Molecular Weight : 265.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9184

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9184
Availability details for
Quantity
  • Availability
  • Add to Cart