COM448684394 Combi-Blocks, Inc.

1-methyl-2-(trifluoromethyl)-1,3-benzodiazole

  • MDL number MFCD00561648
  • Cas Number : 384-46-3
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9196

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9196
Availability details for
Quantity
  • Availability
  • Add to Cart