COM448684399 Combi-Blocks, Inc.

N-Butyl-5-fluoro-2-nitroaniline

  • MDL number MFCD16744222
  • Cas Number : 951666-45-8
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9201

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9201
Availability details for
Quantity
  • Availability
  • Add to Cart