COM448684400 Combi-Blocks, Inc.

2-Bromo-1-fluoro-4-methanesulfonylbenzene

  • MDL number MFCD12032287
  • Cas Number : 959961-65-0
  • Molecular Weight : 253.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9203

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9203
Availability details for
Quantity
  • Availability
  • Add to Cart