COM448684401 Combi-Blocks, Inc.

2-Chloro-1-methanesulfonyl-4-(trifluoromethyl)benzene

  • MDL number MFCD09264544
  • Cas Number : 1000339-68-3
  • Molecular Weight : 258.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9206

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9206
Availability details for
Quantity
  • Availability
  • Add to Cart