COM448684405 Combi-Blocks, Inc.

5-Benzyloxy-N-methyl-2-nitroaniline

  • MDL number MFCD24448841
  • Cas Number : 496837-94-6
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9211

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9211
Availability details for
Quantity
  • Availability
  • Add to Cart