COM448684412 Combi-Blocks, Inc.

Ethyl 7-bromo-1-ethyl-2-methyl-1,3-benzodiazole-5-carboxylate

  • MDL number MFCD24448846
  • Cas Number : 1437794-82-5
  • Molecular Weight : 311.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9219

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9219
Availability details for
Quantity
  • Availability
  • Add to Cart